Learn More
R&D Systems Mouse IL-10 R alpha Recombinant Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Manufacturer: R&D Systems 474MR025/CF
This item is not returnable.
View return policy
Description
The Recombinant Mouse IL-10 R alpha Protein is derived from Sf 21 (baculovirus). The Recombinant Mouse IL-10 R alpha Protein has been validated for the following applications: Bioactivity.Specifications
Lyophilized from a 0.2μm filtered solution in PBS. | |
24 kDa | |
97%, by SDS-PAGE under reducing conditions and visualized by silver stain. | |
Spodoptera frugiperda,Sf 21 (baculovirus)-derived mouse IL-10 R alpha protein Thr24-Asn238 (Lys188Arg and Glu222Gly) | |
Unconjugated |
16154 | |
IL-10R alpha | |
25μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Yes |