Learn More
R&D Systems Mouse IL-11 Recombinant Protein
Manufacturer:  R&D Systems 418ML005/CF
 View more versions of this product
Catalog No. 418ML005CF
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
19 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Yes
16156
IL-11
5μg
Unlabeled