Learn More
R&D Systems Mouse IL-17 RD/SEF Recombinant Protein
Manufacturer:  R&D Systems 2276ML050/CF
 View more versions of this product
Catalog No. 2276ML050CF
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Lyophilized from a 0.2μm filtered solution in PBS.
31.6 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Unlabeled
171463
IL-17 RD/SEF
50μg
Monomer
Yes