Learn More
R&D Systems™ Mouse IL-27 Recombinant Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Supplier: R&D Systems™ 2799ML010
Description
The Recombinant Mouse IL-27 (NS0-expressed) Protein is derived from NS0. The Recombinant Mouse IL-27 (NS0-expressed) Protein has been validated for the following applications: Bioactivity.Specifications
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. | |
48.7 kDa | |
10μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |
246779 | |
IL-27 | |
Mouse myeloma cell line,NS0-derived mouse IL-27 protein, (N-terminus) Mouse EBI3 (Tyr19-Pro228) Accession &Num; O35228 GGGSGGGSGGGSGGGS Mouse p28 (Phe29-Ser234) Accession &Num; Q8K3I6 6-His tag (C-terminus) | |
Unconjugated | |
97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.