Learn More
R&D Systems™ Mouse IL-27 Recombinant Protein Available on GSA/VA Contract for Federal Government customers only.
Manufacturer:  R&D Systems™ 2799ML010/CF
 View more versions of this product
Catalog No. 2799ML010CF
Add to cart
This item is not returnable. View return policy
Description
Description
Measured in an anti-viral assay using HepG2 human hepatocellular carcinoma cells infected with encephalomyocarditis (EMC) virus. Bender H. et al. (2009) Hepatology 50:585. The ED 50 for this effect is 0.6-6 ng/mL.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Supplied as a 0.2μm filtered solution in PBS.
48.7 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Unlabeled
246779
IL-27
10μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Yes