Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Mouse IL-28B/IFN-lambda 3 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
$490.68
Specifications
Gene ID (Entrez) | 338374 |
---|---|
Molecular Weight (g/mol) | 19.7 kDa |
Name | IL-28B/IFN-lambda 3 |
Quantity | 25μg |
Source | E. coli-derived mouse IL-28B/IFN-lambda 3 protein Asp20-Val193,with an N-terminal Met |
Catalog Number | Mfr. No. | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Formulation | Price | Quantity & Availability | |||||
1789ML025
|
R&D Systems™
1789ML025 |
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. |
Each for $490.68
|
|
|||||
1789ML025CF
|
R&D Systems™
1789ML025/CF |
Lyophilized from a 0.2μm filtered solution in PBS. |
Each for $490.68
|
|
|||||
Description
Measured in an anti-viral assay using HepG2 human hepatocellular carcinoma cells infected with encephalomyocarditis (EMC) virus. Sheppard, P. et al. (2003) Nat. Immunol. 4:63. The ED50 for this effect is 0.800-8.00 ng/mL.Specifications
338374 | |
IL-28B/IFN-lambda 3 | |
E. coli-derived mouse IL-28B/IFN-lambda 3 protein Asp20-Val193,with an N-terminal Met | |
Recombinant |
19.7 kDa | |
25μg | |
Unconjugated |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title