Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Mouse Kell Recombinant Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Manufacturer:  R&D Systems™ 1454ZN010

Catalog No. 1454ZN010


Add to cart
This item is not returnable. View return policy

Description

Description

The Recombinant Mouse Kell Protein is derived from NS0. The Recombinant Mouse Kell Protein has been validated for the following applications: Enzyme Activity.
Specifications

Specifications

Supplied as a 0.2 μm filtered solution in Tris, NaCl and ZnCl 2 .
76 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Unlabeled
23925
Kell
10μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening.
Yes
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit