Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Mouse MBL-2 Protein
Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Binding Activity
$490.68
Specifications
Gene ID (Entrez) | 17195 |
---|---|
Molecular Weight (g/mol) | 24 kDa (monomer) |
Name | MBL-2 |
Quantity | 50μg |
Source | Mouse myeloma cell line,NS0-derived mouse MBL-2 protein Glu19-Asp244 |
Catalog Number | Mfr. No. | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Formulation | Price | Quantity & Availability | |||||
2208MB050
|
R&D Systems™
2208MB050 |
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. |
Each for $490.68
|
|
|||||
2208MB050CF
|
R&D Systems™
2208MB050/CF |
Lyophilized from a 0.2μm filtered solution in PBS. |
Each for $490.68
|
|
|||||
Description
The Recombinant Mouse MBL-2 Protein is derived from NS0. The Recombinant Mouse MBL-2 Protein has been validated for the following applications: Binding Activity.Specifications
17195 | |
MBL-2 | |
Mouse myeloma cell line,NS0-derived mouse MBL-2 protein Glu19-Asp244 | |
Oligomer | |
Recombinant |
24 kDa (monomer) | |
50μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title