Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Mouse Noggin (aa 28-232) Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
$503.44
Specifications
Accession Number | P97466 |
---|---|
For Use With (Application) | Bioactivity |
Gene ID (Entrez) | 9241 |
Molecular Weight (g/mol) | M.W. (Observed): 25-35 kDa, reducing conditions; M.W. (theoretical): 23 kDa (monomer) |
Name | Noggin (aa 28-232) |
Catalog Number | Mfr. No. | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Formulation | Price | Quantity & Availability | |||||
6997NG025
|
R&D Systems™
6997NG025 |
Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein. |
Each for $503.44
|
|
|||||
6997NG025CF
|
R&D Systems™
6997NG025/CF |
Lyophilized from a 0.2 μm filtered solution in PBS. |
Each for $503.44
|
|
|||||
Description
Measured by its ability to inhibit BMP-4-induced alkaline phosphatase production by ATDC5 mouse chondrogenic cells. The ED50 for this effect is 0.04-0.16 μg/mL in the presence of 50 ng/mL of Recombinant Human BMP-4 (Catalog # 314-GP ).Specifications
P97466 | |
9241 | |
Noggin (aa 28-232) | |
Mouse myeloma cell line, NS0-derived mouse Noggin protein Gln28-Cys232 | |
<0.01 EU / 1 μg of the protein by the LAL method. | |
Nog | |
Recombinant | |
Noggin |
Bioactivity | |
M.W. (Observed): 25-35 kDa, reducing conditions; M.W. (theoretical): 23 kDa (monomer) | |
25 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, <= -20 °C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, <= -20 °C under sterile conditions after reconstitution. | |
NOG, noggin, SYM1, symphalangism 1 (proximal), synostoses (multiple) syndrome 1, SYNS1, SYNS1A | |
Unconjugated | |
>95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title