Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Mouse PDGF-CC Recombinant Protein

Manufacturer:  R&D Systems™ 1447PC025/CF

 View more versions of this product

Catalog No. 1447PC025CF


Add to cart

Description

Description

Source: E. coli-derived, Val235-Gly345, with an N-terminal Met and a 6-His tag
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 13.4 kDa (monomer)
Formulation: Lyophilized from a 0.2µm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 54635
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
13.4 kDa (monomer)
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Unlabeled
54635
PDGF-CC
25μg
Disulfide-linked homodimer
Yes
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit