Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Mouse Rae-1 epsilon Fc Chimera Recombinant Protein Available on GSA/VA Contract for Federal Government customers only.

Manufacturer:  R&D Systems™ 1135RA050

Catalog No. 1135RA050


Add to cart

Description

Description

Source: Mouse myeloma cell line, NS0-derived, Leu29-Ser231
Purity: 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 49.3 kDa (monomer)
Formulation: Lyophilized from a 0.2µm filtered solution in PBS.
Entrez Gene ID: 379043
Regulatory Status: RUO

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS.
49.3 kDa (monomer)
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
NS0
Disulfide-linked homodimer
Yes
379043
Rae-1 epsilon Fc Chimera
50μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unlabeled
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit