Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems Mouse Serpin A12 Recombinant Enzyme

Manufacturer:  R&D Systems 8338PI025

 View more versions of this product

Catalog No. 83-38P-I025


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

Q7TMF5
68054
Serpin A12
SDS-PAGE: 43-48kDa, reducing conditions
Mouse
RUO
Carrier Protein
Yes
Supplied as a 0.2μm filtered solution in Tris and NaCl
47kDa
95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain at 5μg per lane
25μg
-20 to -70°C
Leu20
Unlabeled

Safety and Handling

ShelfLife : 180 days

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit