Learn More
R&D Systems™ Mouse Slit1 Recombinant Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Manufacturer:  R&D Systems™ 5199SL050
Catalog No. 5199SL050
Add to cart
This item is not returnable. View return policy
Description
Description
The Recombinant Mouse Slit1 Protein is derived from CHO. The Recombinant Mouse Slit1 Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
Bioactivity
97.6 kDa
80%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Chinese Hamster Ovary cell line,CHO-derived mouse Slit1 protein Cys34-Glu900,with a C-terminal 10-His tag
Unconjugated
Lyophilized from a 0.2μm filtered solution in MOPS and NaCl.
Slit1
50μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Yes