Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Mouse TGF-beta 2 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
$381.64
Specifications
Gene ID (Entrez) | 21808 |
---|---|
Molecular Weight (g/mol) | 12.7 kDa |
Name | TGF-beta 2 |
Quantity | 5μg |
Source | Chinese Hamster Ovary cell line,CHO-derived mouse TGF-beta 2 protein Met1-Ser414 |
Catalog Number | Mfr. No. | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Formulation | Price | Quantity & Availability | |||||
7346B2005
|
R&D Systems™
7346B2005 |
Lyophilized from a 0.2μm filtered solution in HCl with BSA as a carrier protein. |
Each for $381.64
|
|
|||||
7346B2005CF
|
R&D Systems™
7346B2005/CF |
Lyophilized from a 0.2μm filtered solution in HCl. |
Each for $381.64
|
|
|||||
Description
The Recombinant Mouse TGF-beta 2 Protein is derived from CHO. The Recombinant Mouse TGF-beta 2 Protein has been validated for the following applications: Bioactivity.Specifications
21808 | |
TGF-beta 2 | |
Chinese Hamster Ovary cell line,CHO-derived mouse TGF-beta 2 protein Met1-Ser414 | |
Disulfide-linked homodimer | |
Recombinant |
12.7 kDa | |
5μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title