Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Mouse Thrombopoietin Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
$451.50 - $1774.00
Specifications
Gene ID (Entrez) | 21832 |
---|---|
Molecular Weight (g/mol) | 35 kDa |
Name | Thrombopoietin/Tpo |
Source | Mouse myeloma cell line,NS0-derived mouse Thrombopoietin/Tpo protein Ser22-Thr356 |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
Catalog Number | Mfr. No. | Formulation | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Formulation | Quantity | Price | Quantity & Availability | ||||
488TO005
|
R&D Systems™
488TO005 |
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. | 5μg |
Each for $451.50
|
|
||||
488TO005CF
|
R&D Systems™
488TO005/CF |
Lyophilized from a 0.2μm filtered solution in PBS. | 5μg |
Each for $451.50
|
|
||||
488TO025
|
R&D Systems™
488TO025 |
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. | 25μg |
Each for $1,223.00
|
|
||||
488TO025CF
|
R&D Systems™
488TO025/CF |
Lyophilized from a 0.2μm filtered solution in PBS. | 25μg |
Each for $1,223.00
|
|
||||
48-8TO-200
|
R&D Systems™
488TO200 |
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. | 200μg |
Each for $1,774.00
|
|
||||
48-8TO-200CF
|
R&D Systems™
488TO200/CF |
Lyophilized from a 0.2μm filtered solution in PBS. | 200μg |
Each for $1,774.00
|
|
||||
Description
The Recombinant Mouse Thrombopoietin Protein is derived from NS0. The Recombinant Mouse Thrombopoietin Protein has been validated for the following applications: Bioactivity.Specifications
21832 | |
Thrombopoietin/Tpo | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
35 kDa | |
Mouse myeloma cell line,NS0-derived mouse Thrombopoietin/Tpo protein Ser22-Thr356 | |
Megakaryocyte colony-stimulating factor, Megakaryocyte growth and development factor, megakaryocyte stimulating factor, MGDF, MGDFC-mpl ligand, MKCSF, MK-CSF, ML, MPL ligand, MPLLG, MPLLGMGC163194, Myeloproliferative leukemia virus oncogene ligand, THCYT1, THPO, thrombopoietin, thrombopoietin nirs variant 1, Tpo, TPOMKCSF | |
Recombinant |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title