Learn More
R&D Systems Mouse VEGF 188 Recombinant Protein
Manufacturer:  R&D Systems 7916MV010
 View more versions of this product
Catalog No. 7916MV010
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Lyophilized from a 0.2μm filtered solution in HCl with BSA as a carrier protein.
22.1 kDa (monomer)
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Unlabeled
22339
VEGF 188
10μg
Disulfide-linked homodimer
Yes