R&D Systems Mouse Wnt-2b Recombinant Protein

Manufacturer: R&D Systems 3900WN025/CF

 View more versions of this product

Catalog No. 39-00W-N025CF

  Check Availability
Add to Cart

    Specifications

    Accession Number AAC25397
    Molecular Weight 38kDa
    Regulatory Status RUO
    Without Additives Carrier Protein
    Recombinant Yes
    Name Wnt-2b
    Purity 95%, by SDS-PAGE with silver staining, under reducing conditions
    Quality Control Testing SDS-PAGE: 41-60kDa, reducing conditions
    Quantity 25μg
    Source Mouse
    Shelf Life 365 days
    Termination Ser57 & Asp71
    Conjugate Unlabeled
    Formulation Lyophilized from a 0.2μm filtered solution in PBS, EDTA and CHAPS
    Gene ID (Entrez) 22414
    Storage Requirements -20 to -70°C
    Preparation Method Reconstitute at 100μg/mL in PBS.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit