Learn More
Tocris Bioscience™ MRS 1477 Available on GSA/VA Contract for Federal Government customers only.
TRPV1 positive allosteric modulator
$163.00 - $683.00
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
524610 Available on GSA/VA Contract for Federal Government customers only. View Documents Tocris Bioscience™
5246/10
Each for $163.00
Add to cart
524650 View Documents Tocris Bioscience™
5246/50
Each for $683.00
Add to cart
Description
Description
TRPV1 positive allosteric modulator. Potentiates vanilloid and proton activation of TRPV1 channels. Exhibits analgesic effects in combination with capsaicin (Cat. No. 0462) in rats. Also prolongs capsaicin-induced hypothermia in mice.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Research Use Only. Not For Human or Veterinary Use