Learn More
R&D Systems MTT Cell Proliferation/Viability Assay
Safe and reliable method minimizes the number of steps necessary to assay for proliferation rate in cultured cells
Manufacturer: R&D Systems 4890025K
Includes: MTT reagent; Detergent Reagent
Description
Tetrazolium compound MTT changes to from yellow to purple
MTT is added directly to the culture medium and is reduced by metabolically active cells to insoluble purple formazan dye crystals
The absorbance of the sample is read directly in the wells at an
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
2500 Tests | |
MTT Cell Proliferation/Viability Assay | |
Minimizes steps to assay for proliferation rate in cultured cells | |
The product requires storage at 2 - 8° C and room temperature. Consult the product insert for specific storage temperatures. Do not use past expiration date. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.