Learn More
R&D Systems NeuroPore Permeabilization and Blocking Reagent
Provides gentle permeabilization and facilitates access to the DNA for the biotinylated dNTPs where CNS tissues will not withstand aggressive protease treatment
Manufacturer: R&D Systems 48206001
This item is not returnable.
View return policy
Description
- Ready-to-use, non-proteolytic
- For use prior to labeling with TACS in situ Apoptosis Detection Kits
- May be used as an antibody diluent in dual labeling experiments detecting both DNA fragmentation and antigens sensitive to Proteinase K treatment
Specifications
2 x 5mL | |
NeuroPore Permeabilization and Blocking Reagent | |
Store the unopened product at 2 - 8° C. Do not use past expiration date. |