Learn More
Tocris Bioscience™ NSC 617145
Werner syndrome helicase (WRN) inhibitor
$162.00 - $666.00
Specifications
Product Type Bioactive Small Molecule
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
534010
View Documents Promotion Details
Tocris Bioscience™
5340/10
10 mg
Each for $162.00
Add to cart
534050
View Documents Promotion Details
Tocris Bioscience™
5340/50
50 mg
Each for $666.00
Add to cart
Description
Description
Werner syndrome helicase (WRN) helicase inhibitor (IC50 = 250 nM); acts synergistically with Mitomycin C (Cat. No. 3258) to induce double-strand breaks and chromosomal abnormalities in vitro.
Specifications
Specifications
Bioactive Small Molecule
Safety and Handling
Safety and Handling
Recommended Storage : Store at -20°C
SDS
Product Certifications
Research Use Only. Not For Human or Veterinary Use