Learn More
R&D Systems™ PARP Enzyme High Specific Activity
Purified from E. coli containing a recombinant plasmid harboring the human PARP gene
Manufacturer:  R&D Systems™ 466850001
 View more versions of this product
Catalog No. 466850001
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
This item is not returnable. View return policy
Description
Description
PARP [Poly(ADP-ribose) Polymerase], also known as ADPRT and PPOL, is a 118kDa enzyme that uses NAD as a substrate to catalyze the covalent transfer of ADP-ribose to a variety of nuclear protein acceptors. ADP ribosyltransferase is required for cellular repair, and PARP expression is induced by single-strand breaks in DNA. PARP is proteolytically cleaved by Caspase-3 into two fragments of 89 and 24kDa in one of the hallmark events of apoptosis.
  • One unit of PARP incorporates 10femtomoles of NAD onto immobilized histones in 30 minutes
  • High specific activity PARP is provided at 10 units/µL
Specifications
Specifications
Poly [ADP-ribose] Polymerase Enzyme High Specific Activity
500μL
Store at -20° C in a manual defrost freezer. For long-term storage, freeze in working aliquots at -80° C. Avoid freeze-thaw cycles.