Learn More
R&D Systems™ Peroxynitrite, Positive Control for Anti-Nitrotyrosine Mab
For use in the preparation of positive control proteins and cell lysates for immunoreactivity assays
Supplier:  R&D Systems™ AR006
Catalog No. AR006
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
This item is not returnable. View return policy
Description
Description
Peroxynitrite is one of the major antimicrobial constituents produced by macrophages. It is usually generated by the nonenzymatic interaction of nitric oxide with superoxide anion. Since it is highly reactive in its own right, Peroxynitrite undergoes addition to a variety of cellular macromolecules, including tyrosine, cysteine, and tryptophan residues in proteins. The tyrosine adducts in proteins are chemically stable and have been used as markers of oxidative damage in pathological conditions such as chronic inflammation, adult respiratory distress syndrome (ARDS) and ischemic brain injury.
  • Synthesized by the reaction of hydrogen peroxide with sodium nitrite under acidic conditions
  • Although highly reactive at physiological pH, Peroxynitrite is stable at highly alkaline pH and can be stored frozen for long periods with minimal decomposition
Specifications
Specifications
Store the unopened product at -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Peroxynitrite
5 mL
Product Suggestions
Product Suggestions