Learn More
R&D Systems Porcine IL-12 Recombinant Protein
Manufacturer:  R&D Systems 912PL025
 View more versions of this product
Catalog No. 912PL025
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Porcine
Unlabeled
IL-12
25μg
Disulfide-linked heterodimer
Yes