Learn More
R&D Systems Porcine IL-6 Recombinant Protein
Manufacturer:  R&D Systems 686PI025
 View more versions of this product
Catalog No. 686PI025
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
21 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Porcine
Yes
399500
IL-6
25μg
Unlabeled