Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems Rat C-Reactive Protein/CRP Recombinant Protein

Manufacturer:  R&D Systems 1744CR200/CF

 View more versions of this product

Catalog No. 1744CR200CF


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS.
24 kDa (monomer)
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Rat
Unlabeled
25419
C-Reactive Protein/CRP
200μg
Homopentamer
Yes
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit