R&D Systems Rat C-Reactive Protein/CRP Recombinant Protein

Manufacturer: R&D Systems 1744CR200

 View more versions of this product

Catalog No. 1744CR200

  Check Availability
Add to Cart

    Specifications

    Molecular Weight 24 kDa (monomer)
    Name C-Reactive Protein/CRP
    Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
    Structural Form Homopentamer
    Gene ID (Entrez) 25419
    Quantity 200μg
    Recombinant Yes
    Source Rat
    Conjugate Unlabeled
    Formulation Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit