Learn More
R&D Systems Rat CX3CL1/Fractalkine Chemokine Domain, aa 25-100 Recombinant Protein
Manufacturer:  R&D Systems 537FT025
 View more versions of this product
Catalog No. 537FT025
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
8.8 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Rat
Yes
89808
CX3CL1/Fractalkine Chemokine Domain, aa 25-100
25μg
Unlabeled