Learn More
R&D Systems Rat CXCL2/GRO beta/MIP-2/CINC-3 Recombinant Protein
Manufacturer:  R&D Systems 525C3025
 View more versions of this product
Catalog No. 525C3025
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
7.6 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Rat
Yes
114105
CXCL2/GRO beta/MIP-2/CINC-3
25μg
Unlabeled