R&D Systems Rat Fas Ligand/TNFSF6 Recombinant Protein

Manufacturer: R&D Systems 1614FL025/CF

 View more versions of this product

Catalog No. 1614FL025CF

  Check Availability
Add to Cart

    Specifications

    Molecular Weight 21 kDa
    Name Fas Ligand/TNFSF6
    Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
    Quantity 25μg
    Recombinant Yes
    Source Rat
    Conjugate Unlabeled
    Formulation Lyophilized from a 0.2μm filtered solution in PBS.
    Gene ID (Entrez) 25385

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit