Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Rat FGF-10 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
$423.40
Specifications
Gene ID (Entrez) | 25443 |
---|---|
Molecular Weight (g/mol) | 20.2 kDa |
Name | FGF-10 |
Quantity | 25μg |
Source | E. coli-derived rat FGF-10 protein Gln37-Ser215,with an N-terminal Met |
Catalog Number | Mfr. No. | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Formulation | Price | Quantity & Availability | |||||
7804FG025
|
R&D Systems™
7804FG025 |
Lyophilized from a 0.2 μm filtered solution in MOPS, Na 2 SO 4 EDTA and DTT with BSA as a carrier protein. |
Each for $423.40
|
|
|||||
7804FG025CF
|
R&D Systems™
7804FG025/CF |
Lyophilized from a 0.2 μm filtered solution in MOPS, Na 2 SO 4 EDTA and DTT. |
Each for $423.40
|
|
|||||
Description
The Recombinant Rat FGF-10 Protein is derived from E. coli. The Recombinant Rat FGF-10 Protein has been validated for the following applications: Bioactivity.Specifications
25443 | |
FGF-10 | |
E. coli-derived rat FGF-10 protein Gln37-Ser215,with an N-terminal Met | |
Monomer | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
20.2 kDa | |
25μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
FGF10, FGF-10, fibroblast growth factor 10, Keratinocyte growth factor 2, KGF2, KGF-2, produced by fibroblasts of urinary bladder lamina propria | |
Recombinant |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title