Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Rat GDNF Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity, Binding Activity
$407.16 - $1276.00
Specifications
Accession Number | Q07731 |
---|---|
For Use With (Application) | Bioactivity, Binding Activity |
Gene ID (Entrez) | 2668 |
Molecular Weight (g/mol) | M.W. (theoretical): 15 kDa (monomer) |
Name | GDNF |
Catalog Number | Mfr. No. | Formulation | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Formulation | Quantity | Price | Quantity & Availability | ||||
512GF010
|
R&D Systems™
512GF010 |
Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein. | 10 μg |
Each for $407.16
|
|
||||
512GF050CF
|
R&D Systems™
512GF050/CF |
Lyophilized from a 0.2 μm filtered solution in PBS. | 50 μg |
Each for $1,276.00
|
|
||||
512GF050
|
R&D Systems™
512GF050 |
Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein. | 50 μg |
Each for $1,276.00
|
|
||||
512GF010CF
|
R&D Systems™
512GF010/CF |
Lyophilized from a 0.2 μm filtered solution in PBS. | 10 μg |
Each for $407.16
|
|
||||
Description
Measured in a cell proliferation assay using SH-SY5Y human neuroblastoma cells. The ED50 for this effect is 2-12 ng/mL in the presence of Recombinant Rat GFR alpha-1/GDNF R alpha-1 Fc Chimera (Catalog # 560-GR ). Measured by its binding ability in a functional ELISA. Immobilized Recombinant Rat GFR alpha-1/GDNF R alpha-1 Fc Chimera (Catalog # 560-GR ) at 1 μg/mL binds Recombinant Rat GDNF with an apparent Kd <4 nM.Specifications
Q07731 | |
2668 | |
GDNF | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. | |
<0.01 EU / 1 μg of the protein by the LAL method. | |
Unconjugated | |
>97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Bioactivity, Binding Activity | |
M.W. (theoretical): 15 kDa (monomer) | |
Spodoptera frugiperda, Sf 21 (baculovirus)-derived rat GDNF protein Ser78-Ile211 | |
Disulfide-linked homodimer | |
Gdnf | |
Recombinant | |
GDNF |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title