Learn More
R&D Systems Rat IL-10 Recombinant Protein
Manufacturer:  R&D Systems 522RL025/CF
 View more versions of this product
Catalog No. 522RL025CF
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Supplied as a 0.2μm filtered solution in NaH2PO4 and NaCl.
19 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Rat
Yes
25325
IL-10
25μg
Unlabeled