Learn More
Tocris Bioscience™ RBC8
RalA and RalB GTPase inhibitor
$261.50 - $992.50
Specifications
Product Type Bioactive Small Molecule
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
541110
View Documents
Tocris Bioscience™
5411/10
10 mg
Each for $261.50
Add to cart
541150
View Documents
Tocris Bioscience™
5411/50
50 mg
Each for $992.50
Add to cart
Description
Description
RalA and RalB GTPase inhibitor (EC50 ∽3.5 μM). Inhibits the binding of Ral to RaLBP1. Exhibits no detectable inhibition of Ras or RhoA activity. Suppresses growth of xenograft tumors in mice. Cell permeable.
Specifications
Specifications
Bioactive Small Molecule
Safety and Handling
Safety and Handling
Recommended Storage : Store at -20°C
SDS
Product Certifications
Research Use Only. Not For Human or Veterinary Use