Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Rhesus Macaque IL-1 beta/IL-1F2 Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

$423.40

Specifications

Molecular Weight (g/mol) 17.3 kDa
Name IL-1 beta/IL-1F2
Quantity 10μg
Source E. coli-derived rhesus macaque IL-1 beta/IL-1F2 protein Ala117-Ser269
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
View More Specs

Products 2
Catalog Number Mfr. No. Formulation Price Quantity & Availability  
Catalog Number Mfr. No. Formulation Price Quantity & Availability  
1318RL010
View Documents
R&D Systems™
1318RL010
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Each for $423.40
Add to Cart
 
1318RL010CF
View Documents
R&D Systems™
1318RL010/CF
Lyophilized from a 0.2μm filtered solution in PBS.
Each for $423.40
Add to Cart
 
Description

Description

The Recombinant Rhesus Macaque IL-1 beta/IL-1F2 Protein is derived from E. coli. The Recombinant Rhesus Macaque IL-1 beta/IL-1F2 Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

17.3 kDa
10μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unconjugated
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
IL-1 beta/IL-1F2
E. coli-derived rhesus macaque IL-1 beta/IL-1F2 protein Ala117-Ser269
catabolin, IL-1, IL1 beta, IL-1 beta, IL1B, IL-1B, IL1-BETA, IL-1F2, IL1F2IL-1 beta, interleukin 1, beta, interleukin-1 beta, preinterleukin 1 beta, pro-interleukin-1-beta
Recombinant
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit