Learn More
R&D Systems Rhesus Macaque IL-1 beta/IL-1F2 Recombinant Protein
Manufacturer:  R&D Systems 1318RL010
 View more versions of this product
Catalog No. 1318RL010
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
IL-1 beta/IL-1F2
10μg
Unlabeled
17.3 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Rhesus Macaque
Yes