R&D Systems TACS Annexin V-FITC Apoptosis Detection Kit
Facilitates rapid fluorometric quantitation of apoptotic cells
Manufacturer: R&D Systems 4830250K
 View more versions of this product
Catalog No. 4830250K
  Check Availability
Add to Cart
    Specifications
    Kit Contents TACS Annexin V-FITC, 250μL; 10X Binding Buffer, 20mL; Propidium Iodide, 2.5mL
    Quantity 250 Tests
    Type TACS Annexin V-FITC Apoptosis Detection Kit
    For Use With (Application) Facilitates rapid fluorometric quantitation of apoptotic cells
  • Description & Specifications
Related Products
Product Content Feedback