Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ TACS TdT In Situ Apoptosis Detection Kit, Blue Label
A TUNEL assay designed for fixed cells, embedded tissue, and frozen tissue using light and electron microscopy
$562.60
Specifications
Quantity | 30 Tests |
---|---|
Product Type | TACS TdT In Situ Apoptosis Detection Kit |
Content And Storage | The product requires storage at -20 to -70° C, 2 - 8° C, and room temperature. Consult the product insert for specific storage temperatures. Do not use past expiration date. |
Includes: Proteinase K Solution; Cytonin™; TdT enzyme, dNTP mix and buffers; Strep-HRP; TACS-Blue Label; Blue Streptavidin-Diluent; Cations; TACS-Nuclease™ and buffer; Nuclear Fast Red
Catalog Number | Mfr. No. | Detection Method | Kit Contents | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Detection Method | Kit Contents | Price | Quantity & Availability | ||||
481130K
|
R&D Systems™
481130K |
Blue Label | Proteinase K, Cytonin, TdT enzyme, dNTPs, StrepHRP, TACS-Blue Label, Blue Streptavidn-Diluent, Cations, TACS-Nuclease, Nuclear Fast Red |
Each for $562.60
|
|
||||
481030K
|
R&D Systems™
481030K |
DAB | TUNEL for fixed cells, embedded tissue, frozen tissue using light and electron microscopy; Reagents to detect DNA fragmentn in situ |
Item Discontinued
This item has been discontinued by the supplier. Please Sign In to view product availability in your area. View alternate product |
N/A | ||||
Description
Contains a highly purified form of the TdT enzyme for the enzymatic incorporation of biotinylated nucleotides
Biotin labeling is achieved using streptavidin-horseradish peroxidase and the colorimetric substrate TACS Blue Label
Specifications
30 Tests | |
TACS TdT In Situ Apoptosis Detection Kit | |
The product requires storage at -20 to -70° C, 2 - 8° C, and room temperature. Consult the product insert for specific storage temperatures. Do not use past expiration date. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title