Learn More
Tocris Bioscience™ TC-G 1008 Available on GSA/VA Contract for Federal Government customers only.
Potent and selective GPR39 agonist
$216.00 - $909.00
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
535550 View Documents Tocris Bioscience™
5355/50
Each for $909.00
Add to cart
535510 Available on GSA/VA Contract for Federal Government customers only. View Documents Tocris Bioscience™
5355/10
Each for $216.00
Add to cart
Description
Description
Potent and selective GPR39 agonist (EC50 values are 0.4 and 0.8 nM for rat and human receptors respectively). Selective over a panel of kinases (IC50s > 10 μM) and displays minimal binding affinity for ghrelin and neurotensin-1 receptors (IC50s > 30 μM). Increases GLP-1 levels in vitro and in vivo. Orally bioavailable.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Research Use Only. Not For Human or Veterinary Use