Learn More
Tocris Bioscience™ TC-I 2014
Potent TRPM8 antagonist
$265.88 - $954.50
Specifications
Product Type Bioactive Small Molecule
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
541010
View Documents
Tocris Bioscience™
5410/10
10 mg
Each for $265.88
Add to cart
541050
View Documents
Tocris Bioscience™
5410/50
50 mg
Each for $954.50
Add to cart
Description
Description
Potent TRPM8 antagonist (IC50 values are 0.8, 3.0 and 4.4 nM for canine, human and rat channels respectively). Exhibits antiallodynic properties in pain models in vivo.
Specifications
Specifications
Bioactive Small Molecule
Safety and Handling
Safety and Handling
Recommended Storage : Store at 4°C
SDS
Product Certifications
Research Use Only. Not For Human or Veterinary Use