Learn More
Tocris Bioscience™ VU 0422288
Selective group III mGlu positive allosteric modulator
$217.00 - $905.50
Specifications
Product Type Bioactive Small Molecule
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
537810
View Documents Promotion Details
Tocris Bioscience™
5378/10
10 mg
Each for $217.00
Add to cart
537850
View Documents Promotion Details
Tocris Bioscience™
5378/50
50 mg
Each for $905.50
Add to cart
Description
Description
Group III mGlu receptor positive allosteric modulator (EC50 values are 108, 125 and 146 nM for mGlu4, mGlu8 and mGlu7 respectively). Exhibits selectivity over a panel of 68 other GPCRs, ion channels and transporters. Potentiates mGlu7-mediated reductions in excitatory post synaptic potentials in hippocampal slices.
Specifications
Specifications
Bioactive Small Molecule
Safety and Handling
Safety and Handling
Recommended Storage : Store at 4°C
SDS
Product Certifications
Research Use Only. Not For Human or Veterinary Use