Learn More
Tocris Bioscience™ VU 0469650 Hydrochloride Available on GSA/VA Contract for Federal Government customers only.
Potent and selective negative allosteric modulator of mGlu1
$201.00 - $844.00
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
537910 Available on GSA/VA Contract for Federal Government customers only. View Documents Tocris Bioscience™
5379/10
Each for $201.00
Add to cart
537950 View Documents Tocris Bioscience™
5379/50
Each for $844.00
Add to cart
Description
Description
Potent and selective negative allosteric modulator of mGlu1 (IC50 = 99 nM). Exhibits >100-fold selectivity for mGlu1 over mGlu2-8 and 68 other GPCRs, ion channels, kinases and transporters. Brain penetrant.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Research Use Only. Not For Human or Veterinary Use