Learn More
Tocris Bioscience™ XD 14
BET bromodomain inhibitor
Specifications
Product Type Bioactive Small Molecule
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
548910
View Documents
Tocris Bioscience™
5489/10
10 mg This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
548950
View Documents
Tocris Bioscience™
5489/50
50 mg This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description
Description
BET bromodomain inhibitor (Kd values are 160, 170, 380, 490, 830 and 850 nM for BRD4(1), BRD2(1), BRD3(1), BRD3(2), BRD2(2) and BRD4(2) respectively). Also inhibits BRDT(1), CBP, p300 and BRDT(2) (Kd values are 1.2, 1.6, 2.6 and 3.7 μM respectively). Attenuates proliferation of HL-60 leukemia cells in vitro.
Specifications
Specifications
Bioactive Small Molecule
Safety and Handling
Safety and Handling
Recommended Storage : Store at -20°C
SDS
Product Certifications
Research Use Only. Not For Human or Veterinary Use