Learn More
R&D Systems™ Z-Leu-Leu-Glu-AMC
Fluorogenic substrate for measuring the chymotrypsin-like peptidylglutamyl peptide-hydrolyzing activity of the 20S proteasome
Supplier:  R&D Systems™ S23005M
Catalog No. S23005M
Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.
This item is not returnable. View return policy
Description
Description
  • Purity >95% by TLC, HPLC
  • Structure confirmed by NMR
Specifications
Specifications
Z-LLE-AMC, C35H44N4O9
Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Recommendations
Recommendations