Learn More
Tocris Bioscience™ Zosuquidar Trihydrochloride
High affinity P-gp inhibitor
$220.00 - $891.00
Specifications
Product Type Bioactive Small Molecule
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
545610
View Documents
Tocris Bioscience™
5456/10
10 mg
Each for $220.00
Add to cart
545650
View Documents
Tocris Bioscience™
5456/50
50 mg
Each for $891.00
Add to cart
Description
Description
High affinity P-glycoprotein (P-gp) inhibitor (Kd = 79 nM). Restores doxorubicin (Cat. No. 2252) sensitivity in P-gp-expressing multidrug (MDR) resistant cancer cell lines. Also potentiates antitumor efficacy of taxol (Cat. No. 1097) in a MDR human non-small cell lung carcinoma xenograft mouse model.
Specifications
Specifications
Bioactive Small Molecule
Safety and Handling
Safety and Handling
Recommended Storage : Store at -20°C
SDS
Product Certifications
Research Use Only. Not For Human or Veterinary Use