Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-Di-2-naphthylprolinol, 99%, Acros Organics

Click to view available options
Quantity:
1g
130798-48-0
C25H23NO
353.465
LNUDNNFEXRHHGY-XMMPIXPASA-N
r-di-2-naphthylprolinol,r-di naphthalen-2-yl pyrrolidin-2-yl methanol,bis naphthalen-2-yl 2r-pyrrolidin-2-ylmethanol,r-+-di-2-naphthyl-2-pyrrolidinemethanol,r-+-lmdi 2-naphthyl-2-pyrrolidinemethanol,bis naphthalen-2-yl 2r-pyrrolidin-2-yl methanol,bis naphthalen-2-yl 2r-pyrrolidin-2-yl methanol,pubchem7066,di 2-naphthyl-d-prolinol,s---alpha,alpha-di 2-naphthyl-2-pyrrolidinemethanol
7020307
dinaphthalen-2-yl-[(2R)-pyrrolidin-2-yl]methanol
C1CC(NC1)C(C2=CC3=CC=CC=C3C=C2)(C4=CC5=CC=CC=C5C=C4)O
130798-48-0
C25H23NO
353.465
LNUDNNFEXRHHGY-XMMPIXPASA-N
r-di-2-naphthylprolinol,r-di naphthalen-2-yl pyrrolidin-2-yl methanol,bis naphthalen-2-yl 2r-pyrrolidin-2-ylmethanol,r-+-di-2-naphthyl-2-pyrrolidinemethanol,r-+-lmdi 2-naphthyl-2-pyrrolidinemethanol,bis naphthalen-2-yl 2r-pyrrolidin-2-yl methanol,bis naphthalen-2-yl 2r-pyrrolidin-2-yl methanol,pubchem7066,di 2-naphthyl-d-prolinol,s---alpha,alpha-di 2-naphthyl-2-pyrrolidinemethanol
7020307
dinaphthalen-2-yl-[(2R)-pyrrolidin-2-yl]methanol
C1CC(NC1)C(C2=CC3=CC=CC=C3C=C2)(C4=CC5=CC=CC=C5C=C4)O
Quantity 1g
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Formula Weight 353.46
Chemical Name or Material (R)-Di-2-naphthylprolinol, 99%