Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-DPN, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
10mg
This item is not returnable. View return policy
524047-78-7
C15H13NO2
239.274
GHZHWDWADLAOIQ-ZDUSSCGKSA-N
r-dpn,unii-bg5cys8sfd,bg5cys8sfd,di-p-hydroxyphenylpropionitrile, r,r-2,3-bis 4-hydroxyphenyl-propionitrile,tocris-1494,benzenepropanenitrile, 4-hydroxy-alpha-4-hydroxyphenyl-, alphar,2r-2,3-bis 4-hydroxyphenyl propanenitrile
6604873
(2R)-2,3-bis(4-hydroxyphenyl)propanenitrile
C1=CC(=CC=C1CC(C#N)C2=CC=C(C=C2)O)O
This item is not returnable. View return policy
524047-78-7
C15H13NO2
239.274
GHZHWDWADLAOIQ-ZDUSSCGKSA-N
r-dpn,unii-bg5cys8sfd,bg5cys8sfd,di-p-hydroxyphenylpropionitrile, r,r-2,3-bis 4-hydroxyphenyl-propionitrile,tocris-1494,benzenepropanenitrile, 4-hydroxy-alpha-4-hydroxyphenyl-, alphar,2r-2,3-bis 4-hydroxyphenyl propanenitrile
6604873
(2R)-2,3-bis(4-hydroxyphenyl)propanenitrile
C1=CC(=CC=C1CC(C#N)C2=CC=C(C=C2)O)O

Enantiomer of DPN (Cat. No. 1494)

Enantiomer of DPN (Cat. No. 1494). Displays higher affinity for estrogen receptor (ER) β over ERα (Ki values are 1.82 and 147 nM respectively).

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Quantity 10mg
Formula Weight 239.27
Percent Purity >98%
Chemical Name or Material (R)-DPN

Recommended Storage : Store at -20°C