Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-Glycidyl butyrate, 92-94%, Thermo Scientific™

Catalog No. AC252830100
Click to view available options
10g; Glass bottle

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 60456-26-0
CAS Min % 92.0
CAS Max % 100.0
Density 1.0350g/mL
Boiling Point 90.0°C (19.0 mmHg)
Flash Point 90°C
Infrared Spectrum Authentic
Assay Percent Range 92-94%
Packaging Glass Bottle
Molecular Formula C7H12O3
Refractive Index 1.4270 to 1.4290
Quantity 10g
MDL Number MFCD00075120
Specific Rotation Condition − 30.00 (20.00°C c=neat)
Synonym r-glycidyl butyrate, r---glycidyl butyrate, r-oxiran-2-ylmethyl butyrate, 2r-oxiran-2-yl methyl butanoate, r-glycicyl butanoate, butanoic acid, 2r-oxiranylmethyl ester, butyric acid r-glycidyl ester, 2r-oxiran-2-ylmethyl butanoate, 2r oxiran-2-yl methyl butanoate
Specific Rotation − 30.00
Solubility Information Solubility in water: insoluble in water.
InChI Key YLNSNVGRSIOCEU-ZCFIWIBFSA-N
SMILES CCCC(=O)OCC1CO1
IUPAC Name [(2R)-oxiran-2-yl]methyl butanoate
Molecular Weight (g/mol) 144.17
PubChem CID 2724536
Formula Weight 144.17
Percent Purity 92 to 94%
Chemical Name or Material (R)-Glycidyl butyrate, 92-94%
Show More Show Less
Hazard Category MOISTURE SENSITIVE
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
TSCA TSCA
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.