Learn More
Bio X Cell Isotype Control, Rat IgG1, Anti-Trinitrophenol, 5 MG
Supplier: Bio X Cell BE0290R005MG
The TNP6A7 monoclonal antibody reacts with trinitrophenol (TNP). Since TNP is not expressed by mammals, this antibody is suitable as an isotype-matched control for rat IgG1 antibodies in most in vivo and in vitro applications.
- Isotype-matched control for rat IgG1 antibodies
- Suitable for in vivo and in vitro applications
- Host species: Rat
- Isotype: Rat IgG1, λ
- Conjugation: Unconjugated
- Formulation: PBS, pH 7.0, contains no stabilizers or preservatives
- Endotoxin: ≤1EU/mg (≤0.001EU/μg)
- Purity: ≥95% determined by SDS-PAGE
- Sterility: 0.2 μm filtration
- Production: Purified from cell culture supernatant in an animal-free facility
- Purification: Protein G
- Molecular weight: 150 kDa
- Storage: Store at stock concentration at 4°C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.