Alfa Aesar™ (R)-(+)-Indoline-2-carboxylic acid, 97%

Manufacturer: Alfa Aesar™ H27626MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH27626MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 98167-06-7
    Molecular Formula C9H9NO2
    Formula Weight 163.18
    MDL Number MFCD00792496
    Chemical Name or Material (R)-(+)-Indoline-2-carboxylic acid

    Specifications

    Formula Weight 163.18
    Molecular Formula C9H9NO2
    MDL Number MFCD00792496
    CAS 98167-06-7
    Melting Point ≈177°C
    Quantity 250mg
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H361-H373-H317
    Suspected of damaging fertility or the unborn child.
    May cause damage to organs through prolonged or repeated exposure.
    May cause an allergic skin reaction.
    Health Hazard 3 GHS P Statement
    P261-P280h-P262-P305+P351+P338
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/clothing.
    Do not get in eyes, on skin, or on clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Chemical Name or Material (R)-(+)-Indoline-2-carboxylic acid
    Assay 97%

    GHS H Statement
    H361-H373-H317
    Suspected of damaging fertility or the unborn child.
    May cause damage to organs through prolonged or repeated exposure.
    May cause an allergic skin reaction.

    GHS P Statement
    P261-P280h-P262-P305+P351+P338
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/clothing.
    Do not get in eyes, on skin, or on clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit